(S)-2-Amino-6-dodecanamidohexanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-Lys(Lauroyl)-OH

A1194041-1G A1194041-25G A1194041-5G
76677-206EA 5.31 USD
76677-206 76677-208 76677-210
(S)-2-Amino-6-dodecanamidohexanoic acid 98%
(S)-2-Amino-6-dodecanamidohexanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₆N₂O₃
MW: 328.4901 g/mol
MDL Number: MFCD09954506
CAS Number: 52315-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR