N-(2,4-Dichlorobenzyl)hydroxylamine hydrochloride 95%

Supplier: Ambeed
Warning

A1193832-100MG A1193832-1G A1193832-250MG
76677-198EA 22.28 USD
76677-198 76677-200 76677-202
N-(2,4-Dichlorobenzyl)hydroxylamine hydrochloride 95%
N-(2,4-Dichlorobenzyl)hydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₃NO
MW: 228.5035 g/mol
MDL Number: MFCD32173630
CAS Number: 139460-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR