(2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid 97%

Supplier: Ambeed
Warning

A1193509-100MG A1193509-250MG A1193509-1G A1193509-50MG
76677-196EA 66 USD
76677-196 77596-222 77866-504 77866-505
(2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid 97%
(2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
CAS Number: 321524-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR