(S)-2-Amino-3-(4-hydroxyphenyl)propanamide dihydrochloride 97%

Supplier: Ambeed
Warning

A1193497-25G A1193497-100G
76677-194EA 61 USD
76677-194 77620-734
(S)-2-Amino-3-(4-hydroxyphenyl)propanamide dihydrochloride 97%
(S)-2-Amino-3-(4-hydroxyphenyl)propanamide dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄Cl₂N₂O₂
MDL Number: MFCD32644567
CAS Number: 2442565-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR