Ethyl 2-((1-phenylcyclopropyl)amino)pyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

A1193427-10MG A1193427-1MG A1193427-25MG A1193427-50MG A1193427-5MG
76677-180EA 122 USD
76677-180 76677-182 76677-184 76677-186 76677-188
Ethyl 2-((1-phenylcyclopropyl)amino)pyrimidine-5-carboxylate 98%
Ethyl 2-((1-phenylcyclopropyl)amino)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₂
MDL Number: MFCD28347705
CAS Number: 1598426-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR