5-Bromo-3-(3-bromo-4,5-dihydroxybenzylidene)indolin-2-one 96%

Supplier: Ambeed
Warning

A1193421-10MG A1193421-1MG A1193421-50MG A1193421-5MG
76677-172EA 65 USD
76677-172 76677-174 76677-176 76677-178
5-Bromo-3-(3-bromo-4,5-dihydroxybenzylidene)indolin-2-one 96%
5-Bromo-3-(3-bromo-4,5-dihydroxybenzylidene)indolin-2-one

Precaución: for Research Use Only

Formula: C₁₅H₉Br₂NO₃
MDL Number: MFCD03235520
CAS Number: 486443-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR