Qstatin 98%

Supplier: Ambeed
Warning

Synonyms: 1-((5-Bromothiophen-2-yl)sulphonyl)-1H-pyrazole

A1193413-10MG A1193413-1MG A1193413-25MG A1193413-50MG A1193413-5MG
76677-162EA 38.2 USD
76677-162 76677-164 76677-166 76677-168 76677-170
Qstatin 98%
Qstatin

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O₂S₂
MW: 293.15 g/mol
MDL Number: MFCD05117319
CAS Number: 902688-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR