3-(Prop-1-en-2-yl)picolinonitrile 97% stabilised

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1193224-100MG A1193224-1G A1193224-250MG A1193224-5G
76677-154EA 41 USD
76677-154 76677-156 76677-158 76677-160
3-(Prop-1-en-2-yl)picolinonitrile 97% stabilised
3-(Prop-1-en-2-yl)picolinonitrile

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.1732 g/mol
CAS Number: 1450634-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR