4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-amine)) 98%

Supplier: Ambeed
Warning

A1193211-100MG A1193211-10G A1193211-1G A1193211-250MG A1193211-5G
76677-144EA 14.86 USD
76677-144 76677-146 76677-148 76677-150 76677-152
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-amine)) 98%
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-amine))

Precaución: for Research Use Only

Formula: C₅₀H₄₀N₄
MW: 696.8794 g/mol
CAS Number: 2252187-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR