5'-(4-Formylphenyl)-2',4',6'-trimethyl-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde 98%

Supplier: Ambeed
Warning

A1193209-100MG A1193209-250MG A1193209-1G A1193209-5G
76677-140EA 33.95 USD
76677-140 76677-142 77620-730 77620-732
5'-(4-Formylphenyl)-2',4',6'-trimethyl-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde 98%
5'-(4-Formylphenyl)-2',4',6'-trimethyl-[1,1':3',1''-terphenyl]-4,4''-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₃₀H₂₄O₃
MW: 432.5098 g/mol
MDL Number: MFCD32667012
CAS Number: 2195343-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR