2,4,6-Trimethylbenzene-1,3,5-tricarbonitrile 98%

Supplier: Ambeed
Danger

A1193194-1G A1193194-250MG A1193194-5G
76677-128EA 278 USD
76677-128 76677-130 76677-132
2,4,6-Trimethylbenzene-1,3,5-tricarbonitrile 98%
2,4,6-Trimethylbenzene-1,3,5-tricarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₉N₃
CAS Number: 1206-85-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR