Triphenylene-2,3,6,7,10,11-hexathiol 98%

Supplier: Ambeed
Warning

A1193192-100MG A1193192-50MG
76677-126EA 436 USD
76677-126 77866-499
Triphenylene-2,3,6,7,10,11-hexathiol 98%
Triphenylene-2,3,6,7,10,11-hexathiol

Precaución: for Research Use Only

Formula: C₁₈H₁₂S₆
CAS Number: 100077-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR