1,1'-Bis(4-aminophenyl)-[4,4'-bipyridine]-1,1'-diium chloride 98%

Supplier: Ambeed
Warning

A1193175-100MG A1193175-1G A1193175-250MG A1193175-5G
76677-098EA 33 USD
76677-098 76677-100 76677-102 77620-724
1,1'-Bis(4-aminophenyl)-[4,4'-bipyridine]-1,1'-diium chloride 98%
1,1'-Bis(4-aminophenyl)-[4,4'-bipyridine]-1,1'-diium chloride

Precaución: for Research Use Only

Formula: C₂₂H₂₀Cl₂N₄
MW: 411.3270 g/mol
CAS Number: 222973-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR