4,4'-(2,2-Diphenylethene-1,1-diyl)dibenzaldehyde 98%

Supplier: Ambeed
Warning

A1193163-100MG A1193163-1G A1193163-250MG
76677-082EA 21 USD
76677-082 76677-084 76677-086
4,4'-(2,2-Diphenylethene-1,1-diyl)dibenzaldehyde 98%
4,4'-(2,2-Diphenylethene-1,1-diyl)dibenzaldehyde

Precaución: for Research Use Only

Formula: C₂₈H₂₀O₂
MW: 388.4500 g/mol
CAS Number: 1601465-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR