4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diylbis(ethyne-2,1-diyl))dibenzaldehyde 98%

Supplier: Ambeed
Warning

A1193159-100MG A1193159-1G A1193159-250MG
76677-076EA 66.84 USD
76677-076 76677-078 76677-080
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diylbis(ethyne-2,1-diyl))dibenzaldehyde 98%
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diylbis(ethyne-2,1-diyl))dibenzaldehyde

Precaución: for Research Use Only

Formula: C₂₄H₁₂N₂O₂S
MW: 392.4293 g/mol
MDL Number: MFCD32667042
CAS Number: 1933562-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR