2,7-Bis(4-aminophenyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone 98%

Supplier: Ambeed
Warning

A1193154-100MG A1193154-1G A1193154-250MG
76677-070EA 95 USD
76677-070 76677-072 76677-074
2,7-Bis(4-aminophenyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone 98%
2,7-Bis(4-aminophenyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone

Precaución: for Research Use Only

Formula: C₂₆H₁₆N₄O₄
MW: 448.4297 g/mol
CAS Number: 49546-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR