2,2'-([1,1':4',1''-Terphenyl]-4,4''-diyl)diacetonitrile 98%

Supplier: Ambeed
Danger

A1193149-100MG A1193149-250MG A1193149-1G
76677-066EA 8 USD
76677-066 76677-068 77620-720
2,2'-([1,1':4',1''-Terphenyl]-4,4''-diyl)diacetonitrile 98%
2,2'-([1,1':4',1''-Terphenyl]-4,4''-diyl)diacetonitrile

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₂
MDL Number: MFCD09924359
CAS Number: 1000559-50-1
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR