2,2'-([2,2'-Bipyridine]-5,5'-diyl)diacetonitrile 98%
Supplier: Ambeed
|
Warning
|
A1193148-100MG
A1193148-1G
A1193148-250MG
A1193148-5G
76677-064EA
93
USD
76677-064
77620-718
77866-487
77866-488
2,2'-([2,2'-Bipyridine]-5,5'-diyl)diacetonitrile 98%
2,2'-([2,2'-Bipyridine]-5,5'-diyl)diacetonitrile
Precaución: for Research Use Only
Formula:
C₁₄H₁₀N₄ |
MDL Number:
MFCD28341138 CAS Number: 286380-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...