14-Mercaptotetradecanoic acid 97%

Supplier: Ambeed
Danger

A1193074-100MG A1193074-1G A1193074-250MG A1193074-5G
76677-040EA 65 USD
76677-040 76677-042 76677-044 76677-046
14-Mercaptotetradecanoic acid 97%
14-Mercaptotetradecanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₂S
MW: 260.4359 g/mol
MDL Number: MFCD32644573
CAS Number: 184639-68-7
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR