tert-Butyl 5-((tert-butyldimethylsilyl)oxy)-3-iodo-1H-indazole-1-carboxylate 97%

Supplier: Ambeed
Warning

A1192560-100MG A1192560-250MG
76677-034EA 6 USD
76677-034 76677-036
tert-Butyl 5-((tert-butyldimethylsilyl)oxy)-3-iodo-1H-indazole-1-carboxylate 97%
tert-Butyl 5-((tert-butyldimethylsilyl)oxy)-3-iodo-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₇IN₂O₃Si
MDL Number: MFCD27938791
CAS Number: 1426425-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR