2-Chlorobenzene-1,3,5-triol 97%

Supplier: Ambeed
Warning

A1192522-100MG A1192522-250MG
76677-030EA 101 USD
76677-030 76677-032
2-Chlorobenzene-1,3,5-triol 97%
2-Chlorobenzene-1,3,5-triol

Precaución: for Research Use Only

Formula: C₆H₅ClO₃
MDL Number: MFCD20133692
CAS Number: 84743-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR