tert-Butyl 8-hydroxyoctanoate 98%

Supplier: Ambeed
Warning

A1192183-100MG A1192183-1G A1192183-250MG A1192183-5G
76677-010EA 50 USD
76677-010 76677-012 76677-014 77866-471
tert-Butyl 8-hydroxyoctanoate 98%
tert-Butyl 8-hydroxyoctanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₃
MW: 216.3172 g/mol
MDL Number: MFCD30472716
CAS Number: 401901-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR