2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 98%

Supplier: Ambeed
Warning

Synonyms: Pyrene-2,7-diboronic acid, pinacol ester

A1191951-100MG A1191951-1G A1191951-250MG A1191951-5G
76676-988EA 27 USD
76676-988 76676-990 76676-992 76676-994
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene 98%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene

Precaución: for Research Use Only

Formula: C₂₈H₃₂B₂O₄
MDL Number: MFCD15143601
CAS Number: 688756-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR