(4-(4-Methylthiazol-5-yl)phenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A1191765-100MG A1191765-1G A1191765-250MG A1191765-25G A1191765-5G
76676-978EA 7 USD
76676-978 76676-980 76676-982 76676-984 76676-986
(4-(4-Methylthiazol-5-yl)phenyl)methanamine hydrochloride 97%
(4-(4-Methylthiazol-5-yl)phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂S
MDL Number: MFCD31567510
CAS Number: 2288710-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR