2-Oxo-2-(p-tolylthio)acetic acid 97%

Supplier: Ambeed
Warning

A1191672-100MG A1191672-1G A1191672-250MG
76676-972EA 28.65 USD
76676-972 76676-974 76676-976
2-Oxo-2-(p-tolylthio)acetic acid 97%
2-Oxo-2-(p-tolylthio)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃S
MW: 196.2230 g/mol
MDL Number: MFCD20643729
CAS Number: 106871-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR