1-(Benzo[d][1,2,3]thiadiazol-6-yl)-3-(3,4-dichlorophenyl)urea ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
BTdCPU
A1191670-10MG
A1191670-1MG
A1191670-5MG
76676-966EA
74.27
USD
76676-966
76676-968
76676-970
1-(Benzo[d][1,2,3]thiadiazol-6-yl)-3-(3,4-dichlorophenyl)urea ≥98%
1-(Benzo[d][1,2,3]thiadiazol-6-yl)-3-(3,4-dichlorophenyl)urea
Precaución: for Research Use Only
Formula:
C₁₃H₈Cl₂N₄OS MW: 339.2 g/mol |
MDL Number:
MFCD32004476 CAS Number: 1257423-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...