tert-Butyl 2',6'-dichloro-5,6-dihydro-[4,4'-bipyridine]-1(2H)-carboxylate 97%

Supplier: Ambeed
Danger

A1191592-100MG A1191592-1G A1191592-250MG A1191592-5G
76676-952EA 26.53 USD
76676-952 76676-954 76676-956 76676-958
tert-Butyl 2',6'-dichloro-5,6-dihydro-[4,4'-bipyridine]-1(2H)-carboxylate 97%
tert-Butyl 2',6'-dichloro-5,6-dihydro-[4,4'-bipyridine]-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₈Cl₂N₂O₂
MW: 329.2216 g/mol
MDL Number: MFCD29054417
CAS Number: 1239363-36-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR