3-Fluoro-6-(trifluoromethyl)picolinic acid 95%

Supplier: Ambeed
Warning

A1189822-100MG A1189822-1G A1189822-250MG
76676-946EA 166.57 USD
76676-946 76676-948 76676-950
3-Fluoro-6-(trifluoromethyl)picolinic acid 95%
3-Fluoro-6-(trifluoromethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MW: 209.0978 g/mol
MDL Number: MFCD15478067
CAS Number: 1214365-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR