2,7-Di-tert-butylpyrene-4,5,9,10-tetraone 98%

Supplier: Ambeed
Warning

A1189609-100MG A1189609-1G A1189609-250MG
76676-940EA 25 USD
76676-940 76676-942 76676-944
2,7-Di-tert-butylpyrene-4,5,9,10-tetraone 98%
2,7-Di-tert-butylpyrene-4,5,9,10-tetraone

Precaución: for Research Use Only

Formula: C₂₄H₂₂O₄
MDL Number: MFCD31700782
CAS Number: 190843-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR