S-Benzo[d]thiazol-2-yl 2-(2-aminothiazol-4-yl)-2-(methoxyimino)ethanethioate 98% + (mixture of isomers)

Supplier: Ambeed
Danger

A1189591-100G A1189591-25G A1189591-500G
76676-934EA 25 USD
76676-934 76676-936 76676-938
S-Benzo[d]thiazol-2-yl 2-(2-aminothiazol-4-yl)-2-(methoxyimino)ethanethioate 98% + (mixture of isomers)
S-Benzo[d]thiazol-2-yl 2-(2-aminothiazol-4-yl)-2-(methoxyimino)ethanethioate

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₄O₂S₃
MDL Number: MFCD00129148
CAS Number: 94088-75-2
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR