PLX5622 (free base) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Fluoro-N-((5-fluoro-2-methoxypyridin-3-yl)methyl)-5-((5-methyl-1H-pyrrolo[2,3-b]pyridin-3-yl)methyl)pyridin-2-amine, PLX5622
A1189464-10MG
A1189464-1MG
A1189464-25MG
A1189464-5MG
76676-926EA
94.43
USD
76676-926
76676-928
76676-930
76676-932
PLX5622 (free base) 98%
PLX5622 (free base)
Precaución: for Research Use Only
Formula:
C₂₁H₁₉F₂N₅O MW: 395.41 g/mol |
MDL Number:
MFCD32201039 CAS Number: 1303420-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...