7-Bromohept-1-yne 97%

Supplier: Ambeed
Danger

A1189336-1G A1189336-250MG A1189336-5G
76676-920EA 181.42 USD
76676-920 76676-922 76676-924
7-Bromohept-1-yne 97%
7-Bromohept-1-yne

Precaución: for Research Use Only

Formula: C₇H₁₁Br
MW: 175.0662 g/mol
MDL Number: MFCD20343079
CAS Number: 81216-14-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR