9-(2-Bromoethyl)-9H-carbazol-3-amine 97%

Supplier: Ambeed
Warning

A1189134-100MG A1189134-250MG
76676-914EA 132.62 USD
76676-914 76676-916
9-(2-Bromoethyl)-9H-carbazol-3-amine 97%
9-(2-Bromoethyl)-9H-carbazol-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrN₂
MW: 289.1704 g/mol
CAS Number: 2442597-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR