N,N-Bis(4-methoxyphenyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-amine 97%

Supplier: Ambeed
Warning

A1189124-100MG A1189124-1G A1189124-250MG
76676-908EA 46 USD
76676-908 76676-910 76676-912
N,N-Bis(4-methoxyphenyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-amine 97%
N,N-Bis(4-methoxyphenyl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-amine

Precaución: for Research Use Only

Formula: C₃₀H₃₂BNO₄
CAS Number: 2225806-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR