O-Ethyl S-((2-(trifluoromethyl)pyridin-3-yl)methyl) carbonodithioate 97%

Supplier: Ambeed
Warning

A1188994-100MG A1188994-1G A1188994-250MG
76676-900EA 9 USD
76676-900 76676-902 76676-904
O-Ethyl S-((2-(trifluoromethyl)pyridin-3-yl)methyl) carbonodithioate 97%
O-Ethyl S-((2-(trifluoromethyl)pyridin-3-yl)methyl) carbonodithioate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NOS₂
MW: 281.3177 g/mol
CAS Number: 2107987-89-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR