(R)-3,3'-Bis(4-methoxyphenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A1188923-100MG A1188923-1G A1188923-250MG A1188923-5G
76676-892EA 39 USD
76676-892 76676-894 76676-896 76676-898
(R)-3,3'-Bis(4-methoxyphenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 98%
(R)-3,3'-Bis(4-methoxyphenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

MDL Number: MFCD32671251
CAS Number: 1011465-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR