Tyrphostin AG 30 98%

Supplier: Ambeed
Warning

A1188508-10MG A1188508-1MG A1188508-25MG A1188508-5MG
76676-864EA 95 USD
76676-864 76676-866 76676-868 76676-870
Tyrphostin AG 30 98%
Tyrphostin AG 30

Precaución: for Research Use Only

Formula: C₁₀H₇NO₄
MDL Number: MFCD09739079
CAS Number: 122520-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR