tert-Butyl 4-(6-nitropyridin-2-yl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A1188474-250MG A1188474-1G
76676-860EA 23 USD
76676-860 77866-447
tert-Butyl 4-(6-nitropyridin-2-yl)piperazine-1-carboxylate 97%
tert-Butyl 4-(6-nitropyridin-2-yl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₄O₄
MW: 308.3330 g/mol
CAS Number: 2222404-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR