Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-((ethoxycarbonothioyl)thio)propanoate, 2-[(Ethoxycarbonothioyl)thio]propionic acid Ethyl ester, Ethyl 2-[(ethoxycarbonothioyl)thio]propanoate, 2-[(Ethoxycarbonothioyl)thio]propanoic acid Ethyl ester

A1188458-100MG A1188458-1G A1188458-250MG A1188458-5G
76676-852EA 31.83 USD
76676-852 76676-854 76676-856 76676-858
Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate 98%
Ethyl 2-[(Ethoxycarbonothioyl)thio]propionate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃S₂
MW: 222.32 g/mol
MDL Number: MFCD30474932
CAS Number: 73232-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR