4-Iodobutan-1-ol 97%

Supplier: Ambeed
Danger

A1188354-1G A1188354-5G A1188354-250MG
76676-848EA 119.89 USD
76676-848 76676-850 77599-710
4-Iodobutan-1-ol 97%
4-Iodobutan-1-ol

Precaución: for Research Use Only

Formula: C₄H₉IO
MW: 200.0181 g/mol
MDL Number: MFCD20414983
CAS Number: 3210-08-0
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR