5'-(4-Carboxyphenyl)-2',4',6'-trimethyl-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A1188287-1G A1188287-250MG A1188287-5G
76676-838EA 91 USD
76676-838 76676-840 77620-688
5'-(4-Carboxyphenyl)-2',4',6'-trimethyl-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid 98%
5'-(4-Carboxyphenyl)-2',4',6'-trimethyl-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₃₀H₂₄O₆
CAS Number: 1246562-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR