1,1,2,2-Tetra-p-tolylethene 99%

Supplier: Ambeed
Warning

A1188263-1G A1188263-250MG A1188263-5G
76676-832EA 35 USD
76676-832 76676-834 76676-836
1,1,2,2-Tetra-p-tolylethene 99%
1,1,2,2-Tetra-p-tolylethene

Precaución: for Research Use Only

Formula: C₃₀H₂₈
MDL Number: MFCD31700768
CAS Number: 5831-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR