(S)-3,3'-Bis(4-(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A1188239-100MG A1188239-1G A1188239-250MG
76676-802EA 50.93 USD
76676-802 76676-804 76676-806
(S)-3,3'-Bis(4-(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 98%
(S)-3,3'-Bis(4-(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

CAS Number: 1228600-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR