(R)-3,3'-Bis(4-nitrophenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A1188211-100MG A1188211-1G A1188211-250MG A1188211-5G
76676-776EA 17 USD
76676-776 76676-778 76676-780 77866-444
(R)-3,3'-Bis(4-nitrophenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 98%
(R)-3,3'-Bis(4-nitrophenyl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₃₂H₂₈N₂O₆
CAS Number: 791616-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR