4,4'-(2,2-Diphenylethene-1,1-diyl)diphenol 95%

Supplier: Ambeed
Warning

A1188118-1G A1188118-250MG A1188118-5G
76676-762EA 155.96 USD
76676-762 76676-764 77620-682
4,4'-(2,2-Diphenylethene-1,1-diyl)diphenol 95%
4,4'-(2,2-Diphenylethene-1,1-diyl)diphenol

Precaución: for Research Use Only

Formula: C₂₆H₂₀O₂
MW: 364.4358 g/mol
CAS Number: 919789-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR