4-Cyano-3,5-difluorophenyl 4-(but-3-en-1-yl)benzoate 95%

Supplier: Ambeed
Warning

A1188052-1G A1188052-5G
76676-758EA 14.86 USD
76676-758 76676-760
4-Cyano-3,5-difluorophenyl 4-(but-3-en-1-yl)benzoate 95%
4-Cyano-3,5-difluorophenyl 4-(but-3-en-1-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₈H₁₃F₂NO₂
MW: 313.2981 g/mol
CAS Number: 208528-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR