Dimethyl 4,4'-oxalyldibenzoate 98%

Supplier: Ambeed
Warning

A1187920-100MG A1187920-1G A1187920-5G
76676-750EA 39.26 USD
76676-750 76676-752 76676-754
Dimethyl 4,4'-oxalyldibenzoate 98%
Dimethyl 4,4'-oxalyldibenzoate

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₆
MW: 326.3002 g/mol
MDL Number: MFCD31758395
CAS Number: 66553-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR