2-Amino-3-methylnaphthalene-1,4-dione 98%

Supplier: Ambeed
Warning

A1187189-1G A1187189-250MG
76676-740EA 121 USD
76676-740 76676-742
2-Amino-3-methylnaphthalene-1,4-dione 98%
2-Amino-3-methylnaphthalene-1,4-dione

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MW: 187.1947 g/mol
MDL Number: MFCD00667088
CAS Number: 7427-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR