5H-Benzo[a]benzo[4,5]thieno[3,2-c]carbazole 98%

Supplier: Ambeed
Warning

A1186877-1G A1186877-250MG
76676-736EA 84.88 USD
76676-736 76676-738
5H-Benzo[a]benzo[4,5]thieno[3,2-c]carbazole 98%
5H-Benzo[a]benzo[4,5]thieno[3,2-c]carbazole

Precaución: for Research Use Only

Formula: C₂₂H₁₃NS
MW: 323.4103 g/mol
CAS Number: 1442458-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR