9'-Phenyl-9'H-9,3':6',9''-tercarbazole 98%

Supplier: Ambeed
Warning

A1186876-100MG A1186876-1G A1186876-250MG A1186876-5G
76676-730EA 22 USD
76676-730 76676-732 76676-734 77866-434
9'-Phenyl-9'H-9,3':6',9''-tercarbazole 98%
9'-Phenyl-9'H-9,3':6',9''-tercarbazole

Precaución: for Research Use Only

Formula: C₄₂H₂₇N₃
MDL Number: MFCD22572690
CAS Number: 211685-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR